6-Aminoimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridin-6-amine

070352-1G 070352-5G
101944-372EA 1221.54 USD
101944-372 101944-374
6-Aminoimidazo[1,2-a]pyridine ≥97%
6-Aminoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.1506 g/mol
MDL Number: MFCD06739232
CAS Number: 235106-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR