5-Amino-7-bromo-1H-indazole ≥96%

Supplier: Matrix Scientific

Synonyms: 7-Bromo-1H-indazol-5-amine

070350-1G 070350-5G
101944-364EA 692.31 USD
101944-364 101944-366
5-Amino-7-bromo-1H-indazole ≥96%
5-Amino-7-bromo-1H-indazole

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD08275114
CAS Number: 953411-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR