5,6-Dihydro-2H-pyran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

070348-1G 070348-5G
101944-356EA 1426.15 USD
101944-356 101944-358
5,6-Dihydro-2H-pyran-3-carboxylic acid ≥95%
5,6-Dihydro-2H-pyran-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₈O₃
MDL Number: MFCD11007815
CAS Number: 100313-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR