Methyl tetrahydro-2H-pyran-3-carboxylate ≥96%

Supplier: Matrix Scientific

070347-1G 070347-5G
101944-352EA 1338.46 USD
101944-352 101944-354
Methyl tetrahydro-2H-pyran-3-carboxylate ≥96%
Methyl tetrahydro-2H-pyran-3-carboxylate

Hazard: Irritant

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
MDL Number: MFCD09839011
CAS Number: 18729-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR