3-(Pyrimidin-4-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

070345-1G 070345-5G
101944-344EA 1396.92 USD
101944-344 101944-346
3-(Pyrimidin-4-yl)benzoic acid ≥95%
3-(Pyrimidin-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
MW: 200.2 g/mol
MDL Number: MFCD12402813
CAS Number: 168619-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR