7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

070342-1G 070342-5G
101944-332EA 509.23 USD
101944-332 101944-334
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClN
MDL Number: MFCD03094742
CAS Number: 220247-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR