1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole hydrochloride

070337-1G 070337-5G
101944-312EA 1396.92 USD
101944-312 101944-314
1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride ≥95%
1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₀ClN₃
MW: 159.62 g/mol
MDL Number: MFCD12400786
CAS Number: 1187830-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR