Ethyl-4-chloropyrrolo[1,2-f][1,2,4]triazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

070336-1G 070336-5G
101944-308EA 1978.46 USD
101944-308 101944-310
Ethyl-4-chloropyrrolo[1,2-f][1,2,4]triazine-2-carboxylate ≥95%
Ethyl-4-chloropyrrolo[1,2-f][1,2,4]triazine-2-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MW: 225.64 g/mol
MDL Number: MFCD11044886
CAS Number: 1120214-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR