6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

070334-1G 070334-5G
101944-300EA 2560 USD
101944-300 101944-302
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridine-2-carboxylic acid ≥95%
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₆N₂O₂
MDL Number: MFCD12400784
CAS Number: 1160995-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR