(6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

070333-1G 070333-5G
101944-296EA 1629.23 USD
101944-296 101944-298
(6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2-yl)methanamine hydrochloride ≥95%
(6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClN₃
MDL Number: MFCD12400783
CAS Number: 1187830-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR