6-Chloropyrido[3,2-b]pyrazine

Supplier: Matrix Scientific

070331-500MG 070331-1G
101944-288EA 1396.92 USD
101944-288 101944-290
6-Chloropyrido[3,2-b]pyrazine
6-Chloropyrido[3,2-b]pyrazine

Hazard: Irritant

Formula: C₇H₄ClN₃
MDL Number: MFCD11518952
CAS Number: 68236-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR