2-(Cyclopropylmethoxy)-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

070328-500MG 070328-1G
101944-276EA 524.62 USD
101944-276 101944-278
2-(Cyclopropylmethoxy)-4-fluorophenylboronic acid pinacol ester ≥97%
2-(Cyclopropylmethoxy)-4-fluorophenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₂BFO₃
MDL Number: MFCD13152010
CAS Number: 1185836-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR