2-(Cyclopropylmethoxy)-5-fluorophenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

070327-500MG 070327-1G
101944-272EA 524.62 USD
101944-272 101944-274
2-(Cyclopropylmethoxy)-5-fluorophenylboronic acid pinacol ester ≥98%
2-(Cyclopropylmethoxy)-5-fluorophenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₂BFO₃
MDL Number: MFCD13152009
CAS Number: 1185836-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR