2-(Cyclopropylmethoxy)phenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

070325-500MG 070325-1G
101944-264EA 524.62 USD
101944-264 101944-266
2-(Cyclopropylmethoxy)phenylboronic acid pinacol ester ≥95%
2-(Cyclopropylmethoxy)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₃BO₃
MDL Number: MFCD13152006
CAS Number: 1185836-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR