2-(Benzyloxy)phenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(2-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Benzyloxyphenylboronic acid pinacol ester

070324-1G 070324-5G
101944-260EA 116.92 USD
101944-260 101944-262
2-(Benzyloxy)phenylboronic acid pinacol ester ≥98%
2-(Benzyloxy)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₉H₂₃BO₃
MDL Number: MFCD03001334
CAS Number: 1027757-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR