2-(Cyclohexylmethoxy)-6-methoxyphenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

070323-500MG 070323-1G
101944-256EA 524.62 USD
101944-256 101944-258
2-(Cyclohexylmethoxy)-6-methoxyphenylboronic acid pinacol ester ≥95%
2-(Cyclohexylmethoxy)-6-methoxyphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₂₀H₃₁BO₄
MDL Number: MFCD13152005
CAS Number: 1204580-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR