2-(Cyclobutylmethoxy)-6-methoxyphenylboronic acid pinacol ester ≥98%
Supplier: Matrix Scientific
070322-500MG
070322-1G
101944-252EA
930.77
USD
101944-252
101944-254
2-(Cyclobutylmethoxy)-6-methoxyphenylboronic acid pinacol ester ≥98%
2-(Cyclobutylmethoxy)-6-methoxyphenylboronic acid pinacol ester
Hazard: Irritant
Formula:
C₁₈H₂₇BO₄ |
MDL Number:
MFCD13152004 CAS Number: 1204580-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...