2-(Cyclopropylmethoxy)-6-fluorophenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

070320-500MG 070320-1G
101944-244EA 524.62 USD
101944-244 101944-246
2-(Cyclopropylmethoxy)-6-fluorophenylboronic acid pinacol ester ≥98%
2-(Cyclopropylmethoxy)-6-fluorophenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₂BFO₃
MDL Number: MFCD13152002
CAS Number: 1204580-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR