2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

070319-500MG 070319-1G
101944-240EA 524.62 USD
101944-240 101944-242
2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester ≥98%
2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₇H₂₅BO₄
MW: 304.2 g/mol
MDL Number: MFCD13152001
CAS Number: 1204580-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR