Methyl-3-iodo-1H-indazole-6-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-iodo-1H-indazole-6-carboxylate

070318-1G 070318-5G
101944-236EA 376.92 USD
101944-236 101944-238
Methyl-3-iodo-1H-indazole-6-carboxylate ≥97%
Methyl 3-iodo-1H-indazole-6-carboxylate

Hazard: Irritant

Formula: C₉H₇IN₂O₂
MW: 302.0686 g/mol
MDL Number: MFCD07781616
CAS Number: 885518-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR