1-Methylpyrrolidin-3-one ≥97%

Supplier: Matrix Scientific

070315-1G 070315-5G
101944-220EA 453.85 USD
101944-220 101944-222
1-Methylpyrrolidin-3-one ≥97%
1-Methylpyrrolidin-3-one

Hazard: Irritant

Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD09991706
CAS Number: 68165-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR