3-Benzyloxypyridine ≥95%
Supplier: Matrix Scientific
070313-1G
070313-5G
101944-210EA
661.54
USD
101944-210
101944-212
3-Benzyloxypyridine ≥95%
3-Benzyloxypyridine
Hazard: Irritant
Formula:
C₁₂H₁₁NO |
MDL Number:
MFCD13151998 CAS Number: 76509-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...