6-Bromo-1,4-dioxaspiro[4.6]undecane ≥98%

Supplier: Matrix Scientific

070310-1G 070310-5G
101944-198EA 1221.54 USD
101944-198 101944-200
6-Bromo-1,4-dioxaspiro[4.6]undecane ≥98%
6-Bromo-1,4-dioxaspiro[4.6]undecane

Hazard: Irritant

Formula: C₉H₁₅BrO₂
MDL Number: MFCD13151996
CAS Number: 70562-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR