2-(1-Bromoethyl)-2-ethyl-1,3-dioxolane ≥98%

Supplier: Matrix Scientific

070309-1G 070309-5G
101944-194EA 930.77 USD
101944-194 101944-196
2-(1-Bromoethyl)-2-ethyl-1,3-dioxolane ≥98%
2-(1-Bromoethyl)-2-ethyl-1,3-dioxolane

Hazard: Irritant

Formula: C₇H₁₃BrO₂
MDL Number: MFCD13151995
CAS Number: 22515-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR