6-Bromo-1,4-dioxaspiro[4.5]decane

Supplier: Matrix Scientific

070308-1G 070308-5G
101944-190EA 930.77 USD
101944-190 101944-192
6-Bromo-1,4-dioxaspiro[4.5]decane
6-Bromo-1,4-dioxaspiro[4.5]decane

Hazard: Irritant

Formula: C₈H₁₃BrO₂
MDL Number: MFCD00086705
CAS Number: 1728-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR