Methyl isoindoline-5-carboxylate ≥95%

Supplier: Matrix Scientific

070306-1G 070306-5G
101944-182EA 1978.46 USD
101944-182 101944-184
Methyl isoindoline-5-carboxylate ≥95%
Methyl isoindoline-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD08234861
CAS Number: 742666-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR