Fmoc-Gln(trt)-Bt ≥98%

Supplier: Matrix Scientific

070303-250MG 070303-500MG
101944-168EA 1106.15 USD
101944-168 101944-170
Fmoc-Gln(trt)-Bt ≥98%
Fmoc-Gln(trt)-Bt

Hazard: Irritant

Formula: C₄₅H₃₇N₅O₄
MDL Number: MFCD12912216
CAS Number: 1155875-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR