Fmoc-Ser(tBu)-Bt ≥98%

Supplier: Matrix Scientific

070302-250MG 070302-500MG
101944-164EA 1106.15 USD
101944-164 101944-166
Fmoc-Ser(tBu)-Bt ≥98%
Fmoc-Ser(tBu)-Bt

Hazard: Irritant

Formula: C₂₈H₂₈N₄O₄
MDL Number: MFCD11850164
CAS Number: 1126433-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR