6-Benzyl-3,5,6,7-tetrahydropyrrolo[3,4-d]pyrimidin-4-one ≥95%

Supplier: Matrix Scientific

070298-500MG 070298-1G
101944-148EA 1106.15 USD
101944-148 101944-150
6-Benzyl-3,5,6,7-tetrahydropyrrolo[3,4-d]pyrimidin-4-one ≥95%
6-Benzyl-3,5,6,7-tetrahydropyrrolo[3,4-d]pyrimidin-4-one

Hazard: Irritant

Formula: C₁₃H₁₃N₃O
MDL Number: MFCD11518995
CAS Number: 1092352-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR