4-Chloro-5-fluoro-2-pyridinemethanol ≥98%

Supplier: Matrix Scientific

Synonyms: (4-Chloro-5-fluoropyridin-2-yl)methanol

070297-500MG 070297-1G 070297-5G
101944-142EA 1629.23 USD
101944-142 101944-144 101944-146
4-Chloro-5-fluoro-2-pyridinemethanol ≥98%
4-Chloro-5-fluoro-2-pyridinemethanol

Hazard: Irritant

Formula: C₆H₅ClFNO
MW: 161.56 g/mol
MDL Number: MFCD09953777
CAS Number: 113209-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR