7-(3-Hydroxypropoxy)quinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

070293-1G 070293-5G
101944-122EA 1512.31 USD
101944-122 101944-124
7-(3-Hydroxypropoxy)quinazolin-4(3H)-one ≥95%
7-(3-Hydroxypropoxy)quinazolin-4(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD08443686
CAS Number: 557770-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR