2-(3-Amino-1H-pyrazol-5-yl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-1H-pyrazole-3-acetic acid

070292-1G 070292-5G
101944-118EA 1396.92 USD
101944-118 101944-120
2-(3-Amino-1H-pyrazol-5-yl)acetic acid ≥95%
2-(3-Amino-1H-pyrazol-5-yl)acetic acid

Hazard: Irritant

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
MDL Number: MFCD09991896
CAS Number: 174891-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR