6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid ≥95%
Supplier: Matrix Scientific
070286-1G
070286-5G
101944-100EA
2560
USD
101944-100
101944-102
6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid ≥95%
6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₇N₃O₄ |
MDL Number:
MFCD04973511 CAS Number: 264623-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...