6-Chloropyrido[3,2-d]pyrimidin-4(3H)-one ≥95%
Supplier: Matrix Scientific
070283-1G
070283-5G
101944-088EA
1050.77
USD
101944-088
101944-090
6-Chloropyrido[3,2-d]pyrimidin-4(3H)-one ≥95%
6-Chloropyrido[3,2-d]pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₇H₄ClN₃O |
MDL Number:
MFCD11846203 CAS Number: 171178-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...