2-Benzyl-5-chloro-1,2,3,4-tetrahydro-2,6-naphthyridine ≥95%

Supplier: Matrix Scientific

070281-1G 070281-5G
101944-080EA 1803.08 USD
101944-080 101944-082
2-Benzyl-5-chloro-1,2,3,4-tetrahydro-2,6-naphthyridine ≥95%
2-Benzyl-5-chloro-1,2,3,4-tetrahydro-2,6-naphthyridine

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MDL Number: MFCD11846202
CAS Number: 1104027-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR