6-Benzyl-5,6,7,8-tetrahydro-2,6-naphthyridin-1(2H)-one ≥95%

Supplier: Matrix Scientific

070280-1G 070280-5G
101944-076EA 1687.69 USD
101944-076 101944-078
6-Benzyl-5,6,7,8-tetrahydro-2,6-naphthyridin-1(2H)-one ≥95%
6-Benzyl-5,6,7,8-tetrahydro-2,6-naphthyridin-1(2H)-one

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD06762738
CAS Number: 601514-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR