1,2,4,5,6,7-Hexahydropyrazolo[3,4-c]pyridin-3-one hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridin-3(2H)-one hydrochloride

070279-1G 070279-5G
101944-072EA 1658.46 USD
101944-072 101944-074
1,2,4,5,6,7-Hexahydropyrazolo[3,4-c]pyridin-3-one hydrochloride ≥95%
1,2,4,5,6,7-Hexahydropyrazolo[3,4-c]pyridin-3-one hydrochloride

Hazard: Irritant

Formula: C₆H₁₀ClN₃O
MW: 175.62 g/mol
MDL Number: MFCD12400785
CAS Number: 1187830-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR