5-Bromo-3-methylbenzo[b]thiophene ≥95%

Supplier: Matrix Scientific

070275-1G 070275-5G
101944-056EA 1629.23 USD
101944-056 101944-058
5-Bromo-3-methylbenzo[b]thiophene ≥95%
5-Bromo-3-methylbenzo[b]thiophene

Hazard: Irritant

Formula: C₉H₇BrS
MDL Number: MFCD05664866
CAS Number: 1196-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR