2-(5-Bromobenzo[b]thiophen-3-yl)ethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

070274-1G 070274-5G
101944-052EA 1570.77 USD
101944-052 101944-054
2-(5-Bromobenzo[b]thiophen-3-yl)ethanamine hydrochloride ≥95%
2-(5-Bromobenzo[b]thiophen-3-yl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₁BrClNS
MDL Number: MFCD01663505
CAS Number: 22964-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR