(5-Bromobenzo[b]thiophen-3-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

070273-1G 070273-5G
101944-048EA 1570.77 USD
101944-048 101944-050
(5-Bromobenzo[b]thiophen-3-yl)methanamine hydrochloride ≥95%
(5-Bromobenzo[b]thiophen-3-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₉H₉BrClNS
MDL Number: MFCD10699250
CAS Number: 744985-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR