6,7-Dihydrocyclopenta[b]pyridin-5-one ≥95%

Supplier: Matrix Scientific

070272-1G 070272-5G
101944-044EA 1280 USD
101944-044 101944-046
6,7-Dihydrocyclopenta[b]pyridin-5-one ≥95%
6,7-Dihydrocyclopenta[b]pyridin-5-one

Hazard: Irritant

Formula: C₈H₇NO
MW: 133.15 g/mol
MDL Number: MFCD07371500
CAS Number: 28566-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR