6,7-Dihydro-1-methyl-1H-indol-4(5H)-one ≥95%

Supplier: Matrix Scientific

070271-1G 070271-5G
101944-040EA 1280 USD
101944-040 101944-042
6,7-Dihydro-1-methyl-1H-indol-4(5H)-one ≥95%
6,7-Dihydro-1-methyl-1H-indol-4(5H)-one

Hazard: Irritant

Formula: C₉H₁₁NO
MDL Number: MFCD01444760
CAS Number: 51471-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR