8-Fluorochroman-4-amine hydrochloride ≥95%

Supplier: Matrix Scientific

070270-1G 070270-5G
101944-036EA 1978.46 USD
101944-036 101944-038
8-Fluorochroman-4-amine hydrochloride ≥95%
8-Fluorochroman-4-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFNO
MDL Number: MFCD08544290
CAS Number: 191608-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR