4-(3-Aminophenyl)pyrrolidin-2-one hydrochloride ≥95%

Supplier: Matrix Scientific

070269-1G 070269-5G
101944-032EA 1920 USD
101944-032 101944-034
4-(3-Aminophenyl)pyrrolidin-2-one hydrochloride ≥95%
4-(3-Aminophenyl)pyrrolidin-2-one hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD11846199
CAS Number: 1187830-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR