7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carbonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

070267-1G 070267-5G
101944-024EA 1744.62 USD
101944-024 101944-026
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carbonitrile hydrochloride ≥95%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carbonitrile hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₀ClF₃N₂
MDL Number: MFCD11846197
CAS Number: 1187830-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR