Methyl-7-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

070265-1G 070265-5G
101944-016EA 1687.69 USD
101944-016 101944-018
Methyl-7-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carboxylate hydrochloride ≥95%
Methyl-7-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline-5-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClF₃NO₂
MDL Number: MFCD11973834
CAS Number: 1187830-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR