Methyl isoindoline-4-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

070264-1G 070264-5G
101944-012EA 1803.08 USD
101944-012 101944-014
Methyl isoindoline-4-carboxylate hydrochloride ≥95%
Methyl isoindoline-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD10700133
CAS Number: 127168-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR