1-Chloropyrrolo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-1H-pyrrolo[1,2-a]pyrazine

070262-1G 070262-5G
101944-004EA 1629.23 USD
101944-004 101944-006
1-Chloropyrrolo[1,2-a]pyrazine ≥95%
1-Chloropyrrolo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD12400782
CAS Number: 136927-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR