Ethyl-8-bromo-4-chloroquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 8-bromo-4-chloroquinoline-3-carboxylate, 8-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

070261-1G 070261-5G
101944-000EA 1687.69 USD
101944-000 101944-002
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate ≥95%
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₉BrClNO₂
MW: 314.563 g/mol
MDL Number: MFCD00173385
CAS Number: 206258-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR