Ethyl-7-chloropyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

070260-1G 070260-5G
101943-996EA 1221.54 USD
101943-996 101943-998
Ethyl-7-chloropyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%
Ethyl-7-chloropyrazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MDL Number: MFCD11973833
CAS Number: 43024-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR